Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013940.1 | NP_001013962.1 | 10 - 11 | 1273 | 83 | |
XM_006239695.2 | XP_006239757.1 | 11 - 12 | 1033 | 83 | ||
GenBank mRNA | BC079201.1 | - | 10 - 11 | 1273 | 7131728 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |