Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007748.1 | NP_001007749.1 | 5 - 6 | 523 | 62 | |
XM_008764597.1 | XP_008762819.1 | 2 - 3 | 1048 | 62 | ||
XM_008764598.1 | XP_008762820.1 | 3 - 4 | 933 | 62 | ||
XM_008764599.1 | XP_008762821.1 | 4 - 5 | 798 | 62 | ||
GenBank mRNA | BC076388.1 | - | 5 - 6 | 523 | 7109513 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |