Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183325.2 | NP_899154.2 | 1 - 2 | 102 | 64 | |
GenBank mRNA | BC082065.1 | - | 1 - 2 | 102 | 7132643 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |