Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759461.1 | XP_008757683.1 | 4 - 5 | 493 | 116 | |
XM_008759462.1 | XP_008757684.1 | 4 - 5 | 492 | 116 | ||
XM_008759463.1 | XP_008757685.1 | 4 - 5 | 488 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |