Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728731.3 | XP_002728777.1 | 10 - 11 | 909 | 93 | |
XM_008759256.1 | XP_008757478.1 | 10 - 11 | 901 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |