Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164656.1 | NP_001158128.1 | 4 - 5 | 1585 | 57 | |
GenBank mRNA | DN933897.1 | - | 1 - 2 | 120 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |