Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nomo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108484.1 | NP_001101954.1 | 4 - 5 | 457 | 93 | |
XM_006229108.2 | XP_006229170.1 | 4 - 5 | 472 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |