Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017514.1 | NP_001017514.1 | 1 - 2 | 216 | 79 | |
GenBank mRNA | BC079244.1 | - | 1 - 2 | 216 | 7132791 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |