Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012168.1 | NP_001012168.1 | 4 - 5 | 600 | 91 | |
XM_006229103.2 | XP_006229165.1 | 7 - 8 | 796 | 91 | ||
XM_006229105.2 | XP_006229167.1 | 7 - 8 | 795 | 91 | ||
XM_006229106.2 | XP_006229168.1 | 7 - 8 | 803 | 91 | ||
XM_006229107.2 | XP_006229169.1 | 4 - 5 | 500 | 91 | ||
XM_008759398.1 | XP_008757620.1 | 4 - 5 | 898 | 91 | ||
GenBank mRNA | BC084696.1 | - | 4 - 5 | 600 | 7115051 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |