Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012067.1 | NP_001012067.1 | 1 - 2 | 248 | 74 | |
GenBank mRNA | BC083883.1 | - | 1 - 2 | 248 | 7131596 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |