Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019175.1 | NP_062048.1 | 1 - 2 | 35 | 71 | |
GenBank mRNA | AF016269.1 | - | 1 - 2 | 35 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |