Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106252.1 | NP_001099722.1 | 4 - 5 | 680 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |