Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107111.1 | NP_001100581.1 | 2 - 3 | 218 | 143 | |
XM_006248062.2 | XP_006248124.1 | 2 - 3 | 447 | 143 | ||
XM_006248063.2 | XP_006248125.1 | 2 - 3 | 791 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |