Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246665.1 | NP_001233594.1 | 5 - 6 | 569 | 66 | |
XM_006249328.2 | XP_006249390.1 | 6 - 7 | 731 | 66 | ||
XM_006249329.2 | XP_006249391.1 | 6 - 7 | 701 | 66 | ||
XM_006249330.2 | XP_006249392.1 | 5 - 6 | 646 | 66 | ||
XM_008769264.1 | XP_008767486.1 | 5 - 6 | 653 | 66 | ||
GenBank mRNA | AB606422.1 | - | 5 - 6 | 569 | 66 | |
DY311222.1 | - | 4 - 5 | 522 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |