Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013094.1 | NP_037226.1 | 4 - 5 | 678 | 105 | |
XM_008759499.1 | XP_008757721.1 | 5 - 6 | 846 | 105 | ||
GenBank mRNA | L26043.1 | - | 4 - 5 | 678 | 105 | |
L26044.1 | - | 4 - 5 | 678 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |