Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229462.2 | XP_006229524.1 | 1 - 2 | 324 | 67 | |
XM_006229463.2 | XP_006229525.1 | 1 - 2 | 324 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |