Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007630.1 | NP_001007631.1 | 2 - 3 | 266 | 127 | |
XM_006229499.2 | XP_006229561.1 | 4 - 5 | 571 | 127 | ||
XM_006229500.2 | XP_006229562.1 | 2 - 3 | 251 | 127 | ||
XM_006229502.2 | XP_006229564.1 | 3 - 4 | 394 | 127 | ||
XM_006229504.1 | XP_006229566.1 | 3 - 4 | 405 | 127 | ||
XM_006229507.2 | XP_006229569.1 | 2 - 3 | 262 | 127 | ||
XM_006229508.2 | XP_006229570.1 | 2 - 3 | 257 | 127 | ||
XM_008759611.1 | XP_008757833.1 | 2 - 3 | 1796 | 127 | ||
GenBank mRNA | BC076394.1 | - | 2 - 3 | 266 | 7107784 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |