Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253318.2 | XP_006253380.1 | 8 - 9 | 980 | 66 | |
XM_008771354.1 | XP_008769576.1 | 10 - 11 | 1976 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |