Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009648.2 | NP_001009648.1 | 2 - 3 | 184 | 140 | |
XM_006231121.2 | XP_006231183.1 | 4 - 5 | 491 | 140 | ||
XM_006231122.2 | XP_006231184.1 | 3 - 4 | 406 | 140 | ||
XM_006231123.2 | XP_006231185.1 | 3 - 4 | 228 | 140 | ||
XM_008760257.1 | XP_008758479.1 | 4 - 5 | 1371 | 140 | ||
GenBank mRNA | AB201944.1 | - | 2 - 3 | 184 | 140 | |
AY325192.1 | - | 3 - 4 | 224 | 140 | ||
BC087043.1 | - | 3 - 4 | 282 | 7108337 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |