Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039031.1 | NP_001034120.1 | 14 - 15 | 1620 | 87 | |
XM_006231056.2 | XP_006231118.1 | 14 - 15 | 1506 | 87 | ||
XM_006231057.2 | XP_006231119.2 | 14 - 15 | 1789 | 87 | ||
GenBank mRNA | BC098925.1 | - | 14 - 15 | 1620 | 7112371 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |