Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106375.2 | NP_001099845.2 | 8 - 9 | 1110 | 117 | |
XM_006231274.2 | XP_006231336.1 | 9 - 10 | 1270 | 117 | ||
GenBank mRNA | BC169061.1 | - | 8 - 9 | 1110 | 5621354 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |