Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005907.2 | NP_001005907.1 | 1 - 2 | 100 | 95 | |
NM_001277341.1 | NP_001264270.1 | 1 - 2 | 100 | 95 | ||
NM_001277346.1 | NP_001264275.1 | 1 - 2 | 100 | 95 | ||
GenBank mRNA | CB811907.1 | - | 1 - 2 | 100 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |