Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106317.2 | NP_001099787.1 | 3 - 4 | 972 | 104 | |
XM_006230537.2 | XP_006230599.1 | 4 - 5 | 1072 | 104 | ||
XM_006230538.2 | XP_006230600.1 | 4 - 5 | 1365 | 104 | ||
XM_006230539.2 | XP_006230601.1 | 3 - 4 | 968 | 104 | ||
GenBank mRNA | BC158743.1 | - | 3 - 4 | 972 | 9100666 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |