Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031675.2 | NP_113863.2 | 3 - 4 | 503 | 98 | |
XM_006228687.1 | XP_006228749.1 | 3 - 4 | 542 | 98 | ||
XM_006228688.1 | XP_006228750.1 | 3 - 4 | 542 | 98 | ||
XM_006228689.1 | XP_006228751.1 | 3 - 4 | 542 | 98 | ||
XM_006228690.2 | XP_006228752.1 | 3 - 4 | 821 | 98 | ||
XM_008759185.1 | XP_008757407.1 | 3 - 4 | 542 | 98 | ||
GenBank mRNA | AF190909.1 | - | 3 - 4 | 428 | 98 | |
AY724534.1 | - | 3 - 4 | 472 | 98 | ||
BC061788.1 | - | 3 - 4 | 494 | 5623735 | 98 | |
U19893.1 | - | 3 - 4 | 504 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |