Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017059.2 | NP_058755.2 | 3 - 4 | 326 | 90 | |
GenBank mRNA | AF235993.1 | - | 2 - 3 | 691 | 90 | |
CB586216.1 | - | 3 - 4 | 326 | 90 | ||
S76511.1 | - | 1 - 2 | 131 | 90 | ||
U32098.1 | - | 1 - 2 | 131 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |