Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053760.2 | NP_446212.2 | 4 - 5 | 337 | 100 | |
XM_006249376.2 | XP_006249438.1 | 4 - 5 | 393 | 100 | ||
XM_008769209.1 | XP_008767431.1 | 4 - 5 | 440 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |