Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181080.1 | NP_851594.1 | 3 - 4 | 725 | 142 | |
XM_006249129.2 | XP_006249191.1 | 4 - 5 | 1047 | 142 | ||
GenBank mRNA | AY081950.1 | - | 3 - 4 | 725 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |