Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098793.1 | NP_001092263.1 | 4 - 5 | 433 | 145 | |
GenBank mRNA | BC158549.1 | - | 4 - 5 | 433 | 8361725 | 145 |
BQ194450.1 | - | 2 - 3 | 145 | |||
DY314400.1 | - | 4 - 5 | 433 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |