Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006962.1 | NP_001006963.1 | 3 - 4 | 621 | 104 | |
XM_006251940.2 | XP_006252002.1 | 3 - 4 | 1609 | 104 | ||
XM_006251942.2 | XP_006252004.1 | 3 - 4 | 1609 | 104 | ||
XM_006251943.2 | XP_006252005.1 | 3 - 4 | 1610 | 104 | ||
XM_006251944.2 | XP_006252006.1 | 3 - 4 | 1609 | 104 | ||
GenBank mRNA | BC083766.1 | - | 3 - 4 | 621 | 7193196 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |