Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam72a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081451.1 | NP_001074920.1 | 2 - 3 | 231 | 117 | |
GenBank mRNA | AY513720.1 | - | 2 - 3 | 231 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |