Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017073.3 | NP_058769.4 | 5 - 6 | 728 | 76 | |
GenBank mRNA | BC061559.1 | - | 5 - 6 | 741 | 6918514 | 76 |
BC072694.1 | - | 5 - 6 | 728 | 7103110 | 76 | |
FQ213256.1 | - | 5 - 6 | 739 | 76 | ||
FQ226911.1 | - | 5 - 6 | 751 | 76 | ||
FQ232853.1 | - | 5 - 6 | 789 | 76 | ||
FQ232963.1 | - | 5 - 6 | 740 | 76 | ||
FQ234289.1 | - | 5 - 6 | 741 | 76 | ||
M29579.1 | - | 5 - 6 | 739 | 76 | ||
M91652.1 | - | 5 - 6 | 737 | 76 | ||
X07921.1 | - | 5 - 6 | 739 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |