Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047902.1 | NP_001041367.1 | 9 - 10 | 1123 | 78 | |
XM_008769482.1 | XP_008767704.1 | 7 - 8 | 900 | 78 | ||
XM_008769483.1 | XP_008767705.1 | 7 - 8 | 897 | 78 | ||
XM_008769484.1 | XP_008767706.1 | 7 - 8 | 873 | 78 | ||
GenBank mRNA | BC111404.1 | - | 2 - 3 | 136 | 78 | |
DQ480752.1 | - | 9 - 10 | 1123 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |