Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250155.2 | XP_006250217.1 | 1 - 2 | 108 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |