Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138879.1 | NP_620234.1 | 3 - 4 | 521 | 77 | |
XM_008769608.1 | XP_008767830.1 | 4 - 5 | 903 | 77 | ||
GenBank mRNA | L25527.1 | - | 3 - 4 | 521 | 77 | |
X69981.1 | - | 3 - 4 | 573 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |