Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007728.1 | NP_001007729.1 | 4 - 5 | 706 | 66 | |
GenBank mRNA | BC078916.1 | - | 4 - 5 | 706 | 7096438 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |