Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109069.1 | NP_001102539.1 | 4 - 5 | 1019 | 69 | |
XM_006249791.2 | XP_006249853.1 | 4 - 5 | 1014 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |