Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107191.1 | NP_001100661.1 | 1 - 2 | 38 | 77 | |
XM_008769648.1 | XP_008767870.1 | 1 - 2 | 83 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |