Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008353.2 | NP_001008354.1 | 3 - 4 | 1737 | 102 | |
XM_008762254.1 | XP_008760476.1 | 3 - 4 | 1697 | 102 | ||
GenBank mRNA | BC083863.1 | - | 3 - 4 | 1715 | 7127425 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |