Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011957.1 | NP_001011957.1 | 8 - 9 | 883 | 62 | |
XM_006234831.2 | XP_006234893.1 | 8 - 9 | 915 | 62 | ||
XM_006234832.2 | XP_006234894.1 | 8 - 9 | 770 | 62 | ||
GenBank mRNA | BC084693.1 | - | 8 - 9 | 883 | 7107022 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |