Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fahd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134834.1 | NP_001128306.1 | 2 - 3 | 403 | 63 | |
XM_006234960.2 | XP_006235022.1 | 2 - 3 | 421 | 63 | ||
GenBank mRNA | BC166933.1 | - | 2 - 3 | 403 | 6921785 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |