Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109606.1 | NP_001103076.1 | 6 - 7 | 747 | 92 | |
XM_006234656.2 | XP_006234718.1 | 6 - 7 | 772 | 92 | ||
XM_006234657.2 | XP_006234719.1 | 6 - 7 | 772 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |