Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005556.1 | NP_001005556.1 | 1 - 2 | 228 | 58 | |
GenBank mRNA | BC081869.1 | - | 1 - 2 | 228 | 7106026 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |