Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013077.1 | NP_001013095.1 | 4 - 5 | 708 | 82 | |
GenBank mRNA | BC083759.1 | - | 4 - 5 | 708 | 7192931 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |