Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047089.1 | NP_001040554.1 | 4 - 5 | 424 | 66 | |
XM_008762089.1 | XP_008760311.1 | 3 - 4 | 2335 | 66 | ||
GenBank mRNA | AB121675.1 | - | 4 - 5 | 424 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |