Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013095.1 | NP_001013113.1 | 8 - 9 | 716 | 100 | |
GenBank mRNA | BC086383.1 | - | 8 - 9 | 716 | 7301572 | 100 |
FQ213005.1 | - | 8 - 9 | 757 | 100 | ||
FQ225883.1 | - | 8 - 9 | 749 | 100 | ||
FQ226276.1 | - | 8 - 9 | 748 | 100 | ||
FQ229703.1 | - | 8 - 9 | 747 | 100 | ||
FQ230888.1 | - | 8 - 9 | 742 | 100 | ||
FQ232595.1 | - | 8 - 9 | 746 | 100 | ||
FQ232661.1 | - | 8 - 9 | 746 | 100 | ||
FQ234018.1 | - | 8 - 9 | 750 | 100 | ||
FQ234727.1 | - | 6 - 7 | 598 | 100 | ||
FQ234929.1 | - | 8 - 9 | 749 | 100 | ||
FQ235264.1 | - | 5 - 6 | 530 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |