Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014087.1 | NP_001014109.2 | 8 - 9 | 1067 | 97 | |
XM_008765933.1 | XP_008764155.1 | 8 - 9 | 1250 | 97 | ||
XM_008765934.1 | XP_008764156.1 | 8 - 9 | 1183 | 97 | ||
XM_008765935.1 | XP_008764157.1 | 7 - 8 | 1292 | 97 | ||
XM_008765936.1 | XP_008764158.1 | 8 - 9 | 1250 | 97 | ||
XM_008765937.1 | XP_008764159.1 | 8 - 9 | 1248 | 97 | ||
XM_008765938.1 | XP_008764160.1 | 8 - 9 | 1248 | 97 | ||
GenBank mRNA | BC085333.1 | - | 8 - 9 | 1067 | 7114511 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |