Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108754.1 | NP_001102224.1 | 3 - 4 | 471 | 101 | |
GenBank mRNA | FQ215152.1 | - | 3 - 4 | 337 | 101 | |
FQ226445.1 | - | 3 - 4 | 337 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |