Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144870.1 | NP_001138342.1 | 2 - 3 | 589 | 59 | |
XM_008765968.1 | XP_008764190.1 | 2 - 3 | 394 | 59 | ||
GenBank mRNA | BC088439.1 | - | 2 - 3 | 433 | 59 | |
DY310175.1 | - | 2 - 3 | 589 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |