Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_574081.6 | XP_574081.4 | 1 - 2 | 584 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |