Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138547.2 | NP_612556.1 | 6 - 7 | 718 | 68 | |
GenBank mRNA | BC091123.1 | - | 6 - 7 | 718 | 7372929 | 68 |
D17310.1 | - | 6 - 7 | 906 | 68 | ||
M64393.1 | - | 6 - 7 | 717 | 68 | ||
S57790.1 | - | 6 - 7 | 676 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |